Skip to Content
Merck
All Photos(1)

Documents

219502

Supelco

Cathepsin L Inhibitor, CAA0225

Synonym(s):

Cathepsin L Inhibitor, CAA0225

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H29N3O5
Molecular Weight:
487.55
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥95% (HPLC)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

solubility

DMSO: 100 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

An epoxysuccinyl peptide highly selective for rat liver cathepsin L (IC50 = 1.9 nM), while exhibiting little to no effect on rat liver cathepsin B at concentrations higher than 1000-5000 nM, in vitro. During autophagy, it has been shown through immunoblotting that CAA0225 (10 µg/ml) effectively inhibits the degradation of LC3-II and GABARAP compared to control, in nutrient-deprived HeLa and Huh-7 cells.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Takahashi, K., et al. 2009. Biol. Pharm. Bull.32, 475.
Ueno, T., et al. 2009. Autophagy5, 878.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pin Li et al.
EMBO molecular medicine, 14(1), e14511-e14511 (2021-11-16)
In the course of our studies aiming to discover vascular bed-specific endothelial cell (EC) mitogens, we identified leukemia inhibitory factor (LIF) as a mitogen for bovine choroidal EC (BCE), although LIF has been mainly characterized as an EC growth inhibitor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service