Skip to Content
Merck
All Photos(1)

Key Documents

1.73634

Supelco

Update cable for Move 100

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
€87.40

€87.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

Select a Size

Change View
1 PKG
€87.40

About This Item

UNSPSC Code:
41116116
NACRES:
SE.11

€87.40


Please contact Customer Service for Availability

Request a Bulk OrderRequest more information

technique(s)

photometry: suitable

compatibility

for use with Spectroquant® Move 100

storage temp.

15-25°C

General description

Spectroquant®
Update cable for Spectroquant Move 100 and Move Cl₂ / O₃ / ClO₂ / CyA / pH

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service