Skip to Content
Merck
All Photos(1)

Documents

1.50209

Supelco

LiChrospher® RP-8 ADS LiChroCART® 25-4

pkg of 3 cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115709

packaging

pkg of 3 cartridges

Quality Level

technique(s)

solid phase extraction (SPE): suitable

storage temp.

no temp limit

General description

3 HPLC cartridges

Analysis Note

Pressure: ≤ 5 bar
Theoretical Plates (N/m) (Toluene): ≥ 3000
Symmetry (Toluene): 0.8 - 2.5
Capacity factor (Toluene): ≥ 0.5

Legal Information

LICHROCART is a registered trademark of Merck KGaA, Darmstadt, Germany
LiChrospher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service