Skip to Content
Merck
All Photos(1)

Key Documents

14-804-M

Sigma-Aldrich

Met (Y1248H) Protein, active

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€423.00
250 μG
€3,470.00

€423.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 μG
€423.00
250 μG
€3,470.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€423.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

100
300

recombinant

expressed in baculovirus infected Sf21 cells

description

protein target Met

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... MET(4233)

General description

FUNCTION: SwissProt: P08581 # Receptor for hepatocyte growth factor and scatter factor. Has a tyrosine-protein kinase activity.
SIZE: 1390 amino acids; 155527 Da
SUBUNIT: Heterodimer formed of an alpha chain (50 kDa) and a beta chain (145 kDa) which are disulfide linked. Binds PLXNB1 and GRB2. Interacts with SPSB1, SPSB2 and SPSB4 (By similarity). Interacts with INPP5D/SHIP1. When phosphorylated at Tyr-1356, interacts with INPPL1/SHIP2. Interacts with RANBP9 and RANBP10.
SUBCELLULAR LOCATION: Membrane; Single-pass type I membrane protein.
DISEASE: SwissProt: P08581 # Activation of MET after rearrangement with the TPR gene produces an oncogenic protein. & Defects in MET may be associated with gastric cancer. & Defects in MET are a cause of hepatocellular carcinoma (HCC) [MIM:114550]. & Defects in MET are a cause of hereditary papillary renal carcinoma (HPRC) [MIM:605074]; also known as papillary renal cell carcinoma 2 (RCCP2). HPRC is a form of inherited kidney cancer characterized by a predisposition to develop multiple, bilateral papillary renal tumors. The pattern of inheritance is consistent with autosomal dominant transmission with reduced penetrance. & Genetic variations in MET may be associated with susceptibility to autism type 1B (AUTS1B) [MIM:608636]. Autism is a neurodevelopmental disorder characterized by disturbance in language, perception and socialization. The disorder is classically defined by a triad of limited or absent verbal communication, a lack of reciprocal social interaction or responsiveness, and restricted, stereotypical, and ritualized patterns of interests and behavior.
SIMILARITY: SwissProt: P08581 ## Belongs to the protein kinase superfamily. Tyr protein kinase family. & Contains 3 IPT/TIG domains. & Contains 1 protein kinase domain. & Contains 1 Sema domain.

Packaging

Also available in 10 μg size (cat#14-804) and in bulk (cat# 14-804-K).
Available in 250 μg --call for pricing and availability and reference catalog number 14-804M when ordering the 250 μg size.

Physical properties

50.1 kDa

Storage and Stability

6 months at -70°C

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service