Skip to Content
Merck
All Photos(1)

Documents

1.46769

Millipore

m-Green Agar

ReadyPlate 55mm plate for microbiological Membrane filtration Analysis of Yeast and Mold released through an ISO 17025 accredited Lab.

Synonym(s):

Schaufuss Pottinger Medium

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

Agency

ISO

Quality Level

form

plate (ready-to-use)

shelf life

9 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (55 mm plate (20 plates per pack))

density

1.09 g/cm3

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

General description

ReadyPlate agar plates are stackable specially designed plates that helps increase safety during handling, transport, and incubation. ReadyPlate agar plates are of 55mm diameter. m-Green Agar is a medium used for the detection of yeast and fungi in beverages. The formulation is rich in nutrients, and a low pH of 4.6 ± 0.2 allows excellent fungal growth. Peptone and yeast extract provides nitrogen, vitamins, minerals, and amino acids essential for growth. Dextrose is a fermentable carbohydrate providing carbon and energy. Magnesium sulfate, thiamine, and diastase (a mixture containing amylolytic (starch) enzymes) provide essential ions, minerals, and nutrients. Bromocresol Green is the pH indicator, facilitating visualization and counting of fungal colonies. The colonies are green due to the diffusion of bromocresol green into the colonies

Application

m-Green Agar is a complex medium for cultivation and isolation of a wide range of yeasts and molds in water samples and Beverages.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service