Skip to Content
Merck
All Photos(1)

Key Documents

1.46761

Millipore

Tryptone Glucose Extract Agar

ReadyPlate ready-to-use 55mm plates for microbiological Membrane filtration Analysis of total Viable count release through an ISO 17025 accredited Lab.

Synonym(s):

Tryptone Glucose Meat extract Agar

Sign Into View Organizational & Contract Pricing

Select a Size

20 EA
€40.90

€40.90


Please contact Customer Service for Availability


Select a Size

Change View
20 EA
€40.90

About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

€40.90


Please contact Customer Service for Availability

Agency

ISO

Quality Level

form

plate (ready-to-use)

shelf life

9 mo.

feature

ready-to-use

manufacturer/tradename

ReadyPlate

packaging

pkg of (55 mm plate (20 plates per pack))

technique(s)

cell counting: suitable (total viable count (TVC))

diam.

55 mm

application(s)

filtration
food and beverages
microbiology

storage temp.

15-25°C

suitability

nonselective for

General description

ReadyPlate agar plates are stackable, specially designed plates that help increase safety during handling, transport, and incubation. ReadyPlate agar plates are of 55mm diameter. Tryptone Glucose Extract Agar is a non-selective medium used for the cultivation and enumeration of microorganisms in water and other filterable liquids by the membrane filtration technique. Tryptone and meat extract provides nitrogenous, carbonaceous compounds, long-chain amino acids, vitamin B complex, and other essential growth nutrients. Glucose serves as the energy source.

Application

Tryptone Glucose Extract Agar is recommended for enumeration of bacteria in water and other filterable liquids by the membrane filtration technique. The media finds its application in the Pharmaceutical, environmental, and Food & Beverages industry.

Legal Information

READYPLATE is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

nwg


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service