Skip to Content
Merck
All Photos(1)

Documents

1.46626

Millipore

Tryptic Soy Agar - Ready-to-use Bottles & Tubes

Histidine, Tween®, Lecithin, Sodium thiosulfate, bottle capacity 125 mL, bottle filling volume 100 mL, closure type, blue screw cap with 3 loci, box of 10 bottles

Synonym(s):

Tryptic Soy Agar with LTHTh

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type blue screw cap with 3 loci
ready-to-use

composition

Histidine
Lecithin
Sodium thiosulfate
Tween®

packaging

box of 10 bottles

bottle capacity

125 mL

bottle filling volume

100 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Agar with LTHTh (Tween® 80, Lecithin, Histidine and Sodium Thiosulfate) is a complex nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microbes present in the test sample. The agar in the medium is used as a gelling agent.
Lecithin and Polysorbate 80, are effective against quaternary ammonium compounds, amphoteric surfactants, benzamidines, chlorhexidines and dequadin. Polysorbate 80 inactivates benzyl alcohol, carbanilides, dichlorbenzyl alcohols, benzoic acid, p-hydroxybenzoic acid and its esters, phenols, phenylethyl alcohols and PHB esters. Histidine and Sodium Thiosulfate is effective against formaldehyde and formaldehyde releasing agents.

Application

Tryptic Soy Agar with LTHTh (Tween® 80, Lecithin, Histidine and Sodium Thiosulfate) is used for the isolation and cultivation of fastidious bacteria, yeasts, and molds.
TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service