Skip to Content
Merck
All Photos(3)

Documents

1.46616

Millipore

Tryptic Soy Broth Ready-to-use Bottles

bottle capacity 275 mL, bottle filling volume 135 mL, closure type, white screw cap for wide neck, box of 6 bottles

Synonym(s):

CASO, CASO Broth, Casein Soya Broth, TSB, Tryptic Soy Broth, Tryptone Soy Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106214

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type white screw cap for wide neck
ready-to-use

packaging

box of 6 bottles

bottle capacity

275 mL

bottle filling volume

135 mL

pH

7.3±0.2

application(s)

cosmetics
food and beverages
pharmaceutical
sterility testing

storage temp.

2-25°C

suitability

nonselective for

Looking for similar products? Visit Product Comparison Guide

General description

Tryptic Soy Broth is a complex nutrient rich inhibitor free microbial media designed in compliance with European (EP), Japanese (JP) and United States Pharmacopeia (USP).
The medium is supplemented with peptones from casein and soymeal to provide an enhanced carbon source, which promotes the growth of the microbes present in the test sample.

Application

Tryptic Soy Broth is used for the isolation and cultivation of fastidious bacteria, yeast. TSB is suitable for sterility testing of substances, preparations, and products from pharmaceutical and food industry.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service