Recommended Products
Agency
ISO 16266
16266
feature
ready-to-use
pH
7.2 ( in H2O)
application(s)
bioburden testing
food and beverages
microbiology
suitability
Pseudomonas spp.
Application
Packaging
Storage and Stability
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service