Skip to Content
Merck
All Photos(1)

Key Documents

14-576

Sigma-Aldrich

Histone H2A.X Protein, human recombinant, 100 µg

Recombinant human Histone H2A.X for use as a substrate in in vitro enzymatic reactions, such as phosphorylation at Ser139 by Rsk1 or MSK1.

Synonym(s):

H2AX, Histone H2AX

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€687.00

€687.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
€687.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

€687.00


Please contact Customer Service for Availability

biological source

human

Quality Level

mol wt

Mw ~15 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

General description

Product Source: produced in E.coli
Recombinant human histone H2A.X

Application

For use as a substrate in in vitro enzymatic reactions, such as phosphorylation at Ser139 by Rsk1 or MSK1.

Quality

routinely evaluated by phosphorylation of Histone H2A.X

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service