Skip to Content
Merck
All Photos(1)

Key Documents

1.30115

Millipore

HY-LiTE® 2 Power Supply Unit

Replacement accessory for HY-LiTE® 2 instruments with built-in printer

Synonym(s):

AC Adapter, Wall Wart, Power Adapter

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€143.00

€143.00


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 UNIT
€143.00

About This Item

UNSPSC Code:
39121004
NACRES:
NA.74

€143.00


Please contact Customer Service for Availability

Request more information

product line

HY-LiTE®

manufacturer/tradename

HY-LiTE®

application(s)

food and beverages

compatibility

for use with HY-LiTE® 2 instruments (with built-in printer (i.e. 1301000301))

storage temp.

no temp limit

General description

Replacement or spare power supply unit (PSU) for HY-LiTE® 2 instruments with built-in printer. HY-LiTE® 2 is a luminometer for monitoring hygiene status in a rapid and reliable way in food and beverage production. The PSU connects to the HY-LiTE® 2 instrument through a low-voltage socket for AC power operation.

Application

Replacement or spare power supply accessory for AC mains power operation of compatible HY-LiTE® 2 instruments.

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service