Trimethyl-Histone H3 (Lys27) Peptide, biotin conjugate, Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes.
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Journal of neuroscience methods, 153(1), 154-161 (2005-11-18)
The development of a technique to load functional indicators into living neurons is an ongoing challenge in retinal neurophysiology. In a number of live-cell preparations, fluorescence-based indicators have been of particular importance for investigating ionic concentrations, protein localization, and other
Immortalization of human uterine leiomyoma and myometrial cell lines after induction of telomerase activity: molecular and phenotypic characteristics.
Sara A Carney,Hidetoshi Tahara,Carol D Swartz,John I Risinger,Hong He,Alicia B Moore et al.
Laboratory Investigation; a Journal of Technical Methods and Pathology null
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.