124039
Akt Inhibitor X
InSolution, ≥97%
Synonym(s):
InSolution Akt Inhibitor X, 10-(4ʹ-(N,N-Diethylamino)butyl)-2-chlorophenoxazine, HCl, 4-(2-Chloro-10H-phenoxazin-10-yl)-N,N-diethylbutan-1-amine, HCl, 4-(2-Chloro-10H-phenoxazin-10-yl)-N,N-diethylbutan-1-amine, HCl, 10-(4ʹ-(N,N-Diethylamino)butyl)-2-chlorophenoxazine, HCl
About This Item
Recommended Products
Quality Level
Assay
≥97% (HPLC)
form
liquid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
avoid repeated freeze/thaw cycles
protect from light
shipped in
dry ice
storage temp.
−70°C
SMILES string
ClC(C=C1)=CC2=C1OC3=C(N2CCCCN(CC)CC)C=CC=C3
InChI
1S/C20H25ClN2O.ClH/c1-3-22(4-2)13-7-8-14-23-17-9-5-6-10-19(17)24-20-12-11-16(21)15-18(20)23;/h5-6,9-12,15H,3-4,7-8,13-14H2,1-2H3;1H
InChI key
SVKSJUIYYCQZEC-UHFFFAOYSA-N
General description
Biochem/physiol Actions
Akt
Packaging
Warning
Physical form
Reconstitution
Other Notes
Thimmaiah, K.N., et al. 2005. J. Biol. Chem.280, 31924.
Legal Information
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service