Skip to Content
Merck
All Photos(1)

Key Documents

12-379

Sigma-Aldrich

Acetyl-Histone H4 Peptide

Lys5/8/12/16, biotin conjugate

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€299.00

€299.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€299.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€299.00


Please contact Customer Service for Availability

Product Name

Acetyl-Histone H4 (Lys5, 8, 12, 16) Peptide, biotin conjugate, Acetyl-Histone H4 (Lys5, 8, 12, 16) Peptide, biotin conjugate primarily used in Immunoprecipitation and Affinity Precipitation Assays.

Quality Level

manufacturer/tradename

Upstate®

technique(s)

affinity precipitation assay: suitable
immunoprecipitation (IP): suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

General description

Synthetic peptide, containing amino acids 1-18 of Histone H4 followed by a GSGS linker and biotinylated lysine (SGRGACKGGACKGLGAC-KGGAACKRHGSGSK-biotin).

Quality

Routinely evaluated in a kinase assay using MAPKAP Kinase 2, active (Catalog # 14-216 & 14-337) to phosphorylate the substrate peptide.

Physical form

Lyophilized powder

Storage and Stability

2 years at -20°C

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening, identification, and characterization of mechanistically diverse inhibitors of the Mycobacterium tuberculosis enzyme, pantothenate kinase (CoaA).
Janani Venkatraman,Jyothi Bhat,Suresh M Solapure,Jatheendranath Sandesh,Debasmita Sarkar et al.
Journal of Biomolecular Screening null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service