Skip to Content
Merck
All Photos(1)

Key Documents

1.20589

Millipore

GranuCult® PEMBA (Polymyxin Pyruvate Egg yolk Mannitol Bromothymolblue Agar) (base)

ISO 21871, Selective Medium for B. cereus

Synonym(s):

PEMBA agar (base)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 21871

Quality Level

sterility

non-sterile; Not processed or packaged aseptically

form

medium granules

manufacturer/tradename

GranuCult®

pH range

7.0 - 7.4 (25 °C)

application(s)

food and beverages
pathogen testing

storage temp.

15-25°C

suitability

selective and differential for bacteria: Bacillus Cereus

General description

GranuCult® PEMBA agar is a selective culture medium containing two indicator systems, mannitol and bromothymol blue, and egg yolk. B. cereus does not utilize mannitol and forms precipitates of hydrolyzed lecithin from the egg yolk.
B. cereus grow as turquoise to peacock blue colonies, since the medium is slightly alkalized by its metabolites (bromothymol blue reaction). The yolk reaction (by lecithinase) also leads to a precipitate surrounding the colony, which appears in the color of the agar or similar to the colony. Growth and lecithinase activity are often so strong that the precipitation is already evident before a colony is visible.
Bacillus cereus is identified by colony morphology, color, and egg yolk hydrolysis. Microscopic examination of the cell and spore morphology and the formation of intracellular lipid globules in vegetative cells is a rapid method of confirmation.
Mannitol-positive bacteria form yellow colonies with a yellow zone in the surrounding medium.

Application

GranuCult® PEMBA agar is used for the selective isolation of viable presumptive Bacillus cereus from Food products and animal feeds and the environmental samples in the area of food production and food handling.

Principle

Low concentration of nutrients and the selective inhibitory component polymyxin help to attain selectivity. Although, some Proteus spp. and Gram-positive cocci also grow in the medium. The low peptone level and the addition of the pyruvate improve the egg yolk precipitation and sporulation. Bromothymol blue acts as a pH indicator to detect mannitol utilization. Enzymatic digest of casein provides carbon and nitrogen sources. Sodium chloride maintains the osmotic balance of the medium, while agar is the solidifying agent.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service