Skip to Content
Merck
All Photos(1)

Key Documents

1.15933

Sigma-Aldrich

Carmine (C.I. 75470)

(calcium-aluminium lacquer of carminic acid) for microscopy Certistain®

Synonym(s):

Carmine (C.I. 75470)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H37AlCaO27 · 3H2O
CAS Number:
Molecular Weight:
1118.85
UNSPSC Code:
41116130
EC Index Number:
215-724-4

Quality Level

form

solid

composition

Dye, ≥50% spectrophotometric assay

solubility

practically insoluble 1.3 g/L

bulk density

290 kg/m3

εmax

105-150 at 525-532 nm in DMSO at 0.05 g/L
80-110 at 565-570 nm in DMSO at 0.05 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-30°C

InChI

1S/C22H20O13/c1-4-8-5(2-6(24)9(4)22(33)34)13(25)10-11(15(8)27)16(28)12(18(30)17(10)29)21-20(32)19(31)14(26)7(3-23)35-21/h2,7,14,19-21,23-24,26,28-32H,3H2,1H3,(H,33,34)

InChI key

DGQLVPJVXFOQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Carmine (C.I. 75470) (calcium-aluminium lacquer of carminic acid) for microscopy Certistain®, is a dry dye that is used for the preparation of a staining solution for staining nuclei and for demonstration of glycogen in histological sections of human origin. As a Certistain® dye it is chemically quality-certified according to highest standards and strict specifications. As an IVD product and CE registered, it can be used hassle-free in diagnostics and laboratory accreditations. For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Analysis Note

Identity
- aluminium:
passes test
- Calcium: passes test
- UV/VIS-Spectrum: passes test
Dye content (spectrophotometrically): ≥ 50 % (als C₂₂H₂₀O₁₃)
Absorption maximum λ 1 (Dimethyl sulfoxide): 565 - 570 nm
Absorption maximum λ 2 (Dimethyl sulfoxide): 525 - 532 nm
Spec. Absorptivity A 1%/1cm
- λ1max; 0.05 g/l; DMSO:
80 - 110
- λ2max; 0.05 g/l; DMSO: 105 - 150
TLC-Test: passes test
Water (according to Karl Fischer): ≤ 15 %
Sulfated ash: 10 - 17 %
Suitability for microscopy: passes test

Legal Information

CERTISTAIN is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service