Skip to Content
Merck
All Photos(1)

Documents

1.13311

Millipore

CULTURA mini incubator (230 V)

For incubation of microbiological samples

Synonym(s):

Incubator

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42281526
NACRES:
NB.14

application(s)

food and beverages

compatibility

for use with incubator

storage temp.

15-25°C

General description

CULTURA mini incubator allows easy incubation of microbiological samples like Envirocheck Contact slides and brings a flexible working environment by reducing the necessary workspace. It provides a tray with holes ready for the Envirocheck Contact slide tubes. It is robust, easy-to-clean, and allows various incubation temperatures.

Application

The CULTURA Mini Incubator is used for the incubation of the samples in small in dimension

Features and Benefits

  • Bench-top system
  • Light-weighted

Legal Information

ENVIROCHECK is a trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Disinfection control solutions ensure food safety and regulatory compliance in food & beverage manufacturing.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service