Skip to Content
Merck
All Photos(1)

Key Documents

1.12048

Sigma-Aldrich

Barium titanate (IV)

Barium titanate (IV). CAS No. 12047-27-7, EC Number 234-975-0.

Synonym(s):

Barium titanate (IV), Barium metatitanate

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€87.80
250 G
€102.00

€87.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€87.80
250 G
€102.00

About This Item

Empirical Formula (Hill Notation):
BaO3Ti
CAS Number:
Molecular Weight:
233.19
MDL number:
UNSPSC Code:
12352303
EC Index Number:
234-975-0
NACRES:
NA.22

€87.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

pH

9.6 (25 °C, 20 g/L in H2O, slurry)

mp

1625 °C

density

6.08 g/cm3 at 20 °C

bulk density

1400 kg/m3

storage temp.

no temp limit

SMILES string

[Ba+2].[Ti+4].[O-2].[O-2].[O-2]

InChI

1S/Ba.3O.Ti/q+2;3*-2;+4

InChI key

VKJLWXGJGDEGSO-UHFFFAOYSA-N

Analysis Note

Identity (elemental analysis): conforms

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service