Skip to Content
Merck
All Photos(1)

Documents

1.09193

Supelco

Ferroin solution

redox indicator solution 1/40 mol/l

Synonym(s):

1,10-Phenanthroline iron(II) salt solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352103
NACRES:
NA.21

product name

Ferroin solution, (1,10-phenanthroline iron(II) sulfate) 1/40 mol/l redox indicator

Quality Level

form

liquid

pH

2.5 (20 °C, 50 g/L in H2O)

density

1.01 g/cm3 at 20 °C

storage temp.

15-25°C

Related Categories

Application


  • Application of Cerium (IV) as an Oxidimetric Agent for the Determination of Ethionamide in Pharmaceutical Formulations: This study utilizes Ferroin solution in analytical chemistry for oxidimetric titration, helping in precise medication formulation (Basavaiah et al., 2016).

  • Optoacoustic spectroscopy for real-time monitoring of strongly light-absorbing solutions in applications to analytical chemistry: Ferroin is part of studies in optoacoustic spectroscopy, aimed at improving real-time monitoring of solutions in lab settings (Filimonova et al., 2013).

  • Sensitized extraction spectrophotometric determination of Hg(II) with dithizone after its flotation as ion-associate using iodide and ferroin: This research highlights the use of Ferroin in sensitive mercury detection methods in environmental studies (Hosseini and Hashemi-Moghaddam, 2005).

Analysis Note

Spec. Absorptivity A 1%/1cm (λmax; 0.003 %; buffer pH 7.0): 180 - 200
Application test: conforms

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service