Skip to Content
Merck
All Photos(1)

Documents

1.08489

Sigma-Aldrich

Urease

lyophilized 5 U/mg EC 3.5.1.5

Synonym(s):

Urease Enzyme

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.21

form

solid

pH

6.0-7.0 (20 °C, 5 g/L in H2O, slurry)

bulk density

410 kg/m3

storage temp.

2-8°C

General description

Urease (urea amidohydrolase; EC 3.5.1.5) is a nickel-dependent metalloenzyme that is found in various plants, bacteria, fungi, algae, and even some invertebrates.

Biochem/physiol Actions

Urease catalyzes the hydrolysis of urea to ammonia and CO2. It is implicated in the development of enzymology and heme protein chemistry. Bacterial urease acts as a virulence factor and is involved in the pathogenesis of hepatic coma, peptic ulceration, and pyelonephritis.

Analysis Note

Activity: ≥ 5 U/mg
Suitability for determination of urea: passes test

Pictograms

Exclamation markHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service