Skip to Content
Merck
All Photos(1)

Key Documents

1.08357

Millipore

Triethanolamine hydrochloride

GR for analysis

Synonym(s):

Triethanolamine hydrochloride, 2,2′,2′′-Nitrilitriethanol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€48.50
1 KG
€329.00

€48.50


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
€48.50
1 KG
€329.00

About This Item

Empirical Formula (Hill Notation):
C6H15NO3 · xHCl
CAS Number:
Molecular Weight:
149.19 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
211-284-2
NACRES:
NA.25

€48.50


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

<1 hPa ( 20 °C)

Quality Level

form

solid

pH

4.0-5.5 (20 °C, 50 g/L in H2O)

mp

180.24 °C

density

1.3539 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

[Cl-].[N+H](CCO)(CCO)CCO

InChI

1S/C6H15NO3.ClH/c8-4-1-7(2-5-9)3-6-10;/h8-10H,1-6H2;1H

InChI key

HHLJUSLZGFYWKW-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99 %
Identity (IR-spectrum): passes test
pH-value (5 %; water): 4.0 - 5.5
Melting range: 176 - 180 °C
Cu (Copper): ≤ 0.0005 %
Fe (Iron): ≤ 0.0005 %
Pb (Lead): ≤ 0.0005 %
Zn (Zinc): ≤ 0.0005 %
Sulfated ash: ≤ 0.01 %

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service