Skip to Content
Merck
All Photos(1)

Key Documents

1.07360

Millipore

Pimaricin

for biochemistry

Synonym(s):

Pimaricin, Natamycin

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
665.73
MDL number:
UNSPSC Code:
12352203
EC Index Number:
231-683-5
NACRES:
NA.21

Quality Level

potency

2730 mg/kg LD50, oral (Rat)

mp

>250 °C (decomposition)

solubility

0.1 g/L

bulk density

200 kg/m3

storage temp.

2-8°C

General description

for biochemistry

Application

Pimaricin for biochemistry. CAS 7681-93-8, molar mass 665.73 g/mol.

Analysis Note

Purity (HPLC)≥ 80.0 % (a/a)Identity (IR-spectrum)conformsSpec. Absorptivity A 1%/1cm (λmax; 0.0008 %; 1 cm; N,N-Dimethylformamide; calc.on anhydrous substance)1000 - 1100Water (according to Karl Fischer)≤ 5.0 %

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service