Skip to Content
Merck
All Photos(1)

Key Documents

1.07069

Supelco

Phloroglucinol

(1,3,5-trihydroxybenzene) for analysis

Synonym(s):

Phloroglucinol, 1,3,5-Trihydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€57.00

€57.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€57.00

About This Item

Linear Formula:
C6H3-1,3,5-(OH)3
CAS Number:
Molecular Weight:
126.11
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-611-2
NACRES:
SE.11

€57.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

crystals
fine powder

potency

4000 mg/kg LD50, oral (Rat)

mp

217-220 °C

solubility

10 g/L

bulk density

560 kg/m3

storage temp.

15-25°C

SMILES string

Oc1cc(cc(c1)O)O

InChI

1S/C6H6O3/c7-4-1-5(8)3-6(9)2-4/h1-3,7-9H

InChI key

QCDYQQDYXPDABM-UHFFFAOYSA-N

Application


  • Biomimetic Synthesis and Chemical Proteomics Reveal the Mechanism of Action and Functional Targets of Phloroglucinol Meroterpenoids.: This study explores the synthesis and mechanisms of phloroglucinol-based compounds, focusing on their biological activity and potential therapeutic applications. It highlights the use of chemical proteomics to identify functional targets, contributing to the understanding of phloroglucinol′s role in medicinal chemistry (Bracken et al., 2024).

  • Clinical efficacy and metabolomics study of Wendan Decoction in the treatment of phlegm-dampness obstructive sleep apnea-hypopnea syndrome with type 2 diabetes mellitus.: This research investigates the clinical effects and metabolic pathways influenced by Wendan Decoction, which includes phloroglucinol as a component. The study uses metabolomics to elucidate the biochemical interactions and therapeutic effects in patients with sleep apnea and diabetes (Zhou et al., 2023).

  • Metabolomics characterizes early metabolic changes and markers of tolerant Eucalyptus ssp. clones against drought stress.: This paper employs metabolomics to study drought tolerance in Eucalyptus clones, identifying early metabolic changes involving phloroglucinol derivatives. The findings enhance the understanding of plant resilience mechanisms under stress conditions (Noleto-Dias et al., 2023).

  • Phloroglucinol and Its Derivatives: Antimicrobial Properties toward Microbial Pathogens.: This review compiles data on the antimicrobial activities of phloroglucinol and its derivatives, emphasizing their potential as bioactive agents in combating various microbial pathogens. The study covers the spectrum of their efficacy and possible applications in microbial control (Khan et al., 2022).

  • Anti-Inflammatory Markers of Hops Cultivars (Humulus lupulus L.) Evaluated by Untargeted Metabolomics Strategy.: This research identifies anti-inflammatory compounds in hops, including phloroglucinol derivatives, using an untargeted metabolomics approach. The study provides insights into the biochemical basis of the anti-inflammatory properties of hops (Nicacio et al., 2022).

Analysis Note

Color (visual): white to yellowish
Appearance of substance (visual): fine powder or crystals
Assay (GC, area%): ≥ 99.0 % (a/a)
Loss on Drying (105°C): ≤ 1.0 %
Sulfated ash: ≤ 0.1 %
Diresorcinol: passes test
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service