Skip to Content
Merck
All Photos(4)

Documents

1.07020

Supelco

Dichloromethane

for analysis EMPARTA® ACS

Synonym(s):

DCM, Methylene chloride, Methylene dichloride, Methylene chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CH2Cl2
CAS Number:
Molecular Weight:
84.93
Beilstein:
1730800
MDL number:
UNSPSC Code:
12191502
EC Index Number:
200-838-9
NACRES:
NA.21

grade

ACS reagent

Quality Level

vapor density

2.9 (vs air)

vapor pressure

24.45 psi ( 55 °C)
6.83 psi ( 20 °C)

product line

EMPARTA®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

1223 °F
605 °C (DIN 51794)

potency

1600 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

22 %

impurities

≤0.00002% Free chlorine (as Cl)
≤0.0003 meq/g Titrable acid
≤0.02% Water

evapn. residue

≤0.002%

color

clearAPHA: ≤10

refractive index

n20/D 1.424 (lit.)

pH

7 (20 °C)

bp

39.8-40 °C (lit.)

mp

-95 °C
−95 °C (lit.)

density

1.325 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

ClCCl

InChI

1S/CH2Cl2/c2-1-3/h1H2

InChI key

YMWUJEATGCHHMB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

EMD Millipore offers EMPARTA grade solvents for routine tasks in analytical laboratories. EMPARTA grade solvents offer fewer test parameters than EMSURE, yet are still of high quality. EMPARTA solvents meet ACS requirements, making them ideal for a wide range of analytical applications.

Analysis Note

Evaporation Number: 1.9
Purity (GC): ≥ 99.5 %
Identity (IR): conforms
Appearance: clear
Color: ≤ 10 Hazen
Titrable acid: ≤ 0.0003 meq/g
Free chlorine (as Cl): ≤ 0.00002 %
Evaporation residue: ≤ 0.002 %
Water: ≤ 0.02 %
Stabilized with about 50 ppm 2-Methyl-2-butene.

Legal Information

EMPARTA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service