Skip to Content
Merck

1.05422

SAFC

Ammonia solution

25%, EMPROVE® ESSENTIAL, Ph. Eur., BP

Pharma Manufacturing

Synonym(s):

Ammonium hydroxide solution, Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€36.00
2.5 L
€54.00
2.5 L
€54.00
25 L
€230.00
180 L
Inquire

€36.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€36.00
2.5 L
€54.00
2.5 L
€54.00
25 L
€230.00
180 L
Inquire

About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352402

€36.00


Please contact Customer Service for Availability

Request a Bulk Order

Agency

BP
Ph. Eur.

Quality Level

vapor pressure

483 hPa ( 20 °C)

product line

EMPROVE® ESSENTIAL

form

liquid

expl. lim.

27 %

concentration

25%

bp

37.7 °C/1013 hPa

mp

-57.5 °C

density

0.9 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge in formulation. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Ammonia solution is a pH-adjusting or buffering agent typically used in a variety of liquid formulations.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service