Skip to Content
Merck
All Photos(1)

Documents

1.04604

Supelco

Conductivity water (nominal 0 mS/cm) certified reference material

new

Test solution for measurement of electrolytic conductivity, traceable to PTB

Sign Into View Organizational & Contract Pricing


About This Item

Agency

traceable to PTB

Quality Level

form

liquid

refractive index

n20/D 1.34 (lit.)

pH

7 (20 °C)

bp

100 °C (lit.)

mp

0 °C (lit.)

density

1.000 g/mL at 3.98 °C (lit.)

storage temp.

15-25°C

General description

This certified reference material (CRM)is directly traceable to the corresponding primary reference material from the Physikalisch Technische Bundesanstalt (PTB), Germany. The electrolytic conductivity of this standard is determined using a conductivity meter from an accredited Merck calibration laboratory.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

Intended for use in aqueous solution for measuring conductivity cell constant or as a control sample.

Analysis Note

Electrical conductivity (25 °C): 0 - 0.002 mS/cm

Legal Information

CERTIPUR®
CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service