Skip to Content
Merck
All Photos(1)

Key Documents

1.04157

Sigma-Aldrich

Fluorescein-5-isothiocyanate (FITC)

Sign Into View Organizational & Contract Pricing

Select a Size

1041570250
€428.00

€428.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

Select a Size

Change View
1041570250
€428.00

About This Item

Empirical Formula (Hill Notation):
C21H11NO5S
CAS Number:
Molecular Weight:
389.38
MDL number:
UNSPSC Code:
41116130
NACRES:
NA.21

€428.00


Estimated to ship on28 March 2025Details


Request a Bulk Order

composition

Dye, ≥95% spectrophotometric assay

Quality Level

mp

>360 °C

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

S=C=Nc1cc2c(cc1)C3(OC2=O)c4c(cc(cc4)O)Oc5c3ccc(c5)O

InChI

1S/C21H11NO5S/c23-12-2-5-16-18(8-12)26-19-9-13(24)3-6-17(19)21(16)15-4-1-11(22-10-28)7-14(15)20(25)27-21/h1-9,23-24H

InChI key

MHMNJMPURVTYEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

In immuno histology/ cytology, Fluorescein-5-isothiocyanate (FITC) for biochemistry and for fluorescence microscopy, serve as marker substance for the visualization and for the detection of tissue-bound bio molecules like e.g. antigens, lectins, various proteins, peptides, nucleic acids, oligo- and polysaccharides in samples of human origin. FITC-marked antibodies bind specifically to the appropriate target structures in the tissue. An optimum FITC-protein (antibody) ratio is important for a good result.

Application

for biochemistry and for fluorescence microscopy

Analysis Note

Dye content (spectrophotometrically): ≥ 95 %
Identity (UV/VIS-Spectrum): passes test
Absorption maximum λ max (Sodium hydroxide solution 0.1 mol/l): 488 - 494 nm
Spec. Absorptivity A 1%/1cm (λmax; 0.003 g/l; NaOH 0.1 mol/l): 2200 - 2380
TLC-Test: passes test

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service