Skip to Content
Merck
All Photos(1)

Key Documents

1.04092

Supelco

Glycerol

Synonym(s):

Trihydroxylpropane, Protol, 1,2,3-Propanetriol, Glycerin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
92.09
Beilstein:
635685
MDL number:
UNSPSC Code:
50161901
technique(s):
electrophoresis: suitable
bp:
182 °C/20 mmHg (lit.)
vapor pressure:
<1 mmHg ( 20 °C)
To inquire about this product 1.04092, please contact your local Merck office or dealer. Contact Technical Service

vapor density

3.1 (vs air)

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

form

liquid

autoignition temp.

698 °F

technique(s)

electrophoresis: suitable

refractive index

n20/D 1.474 (lit.)

pH

~5 (20 °C, 100 g/L)

bp

182 °C/20 mmHg (lit.)

mp

20 °C (lit.)

density

1.25 g/mL (lit.)

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

390.2 °F - Pensky-Martens closed cup

Flash Point(C)

199 °C - Pensky-Martens closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

B Tenderis et al.
International journal of food microbiology, 321, 108560-108560 (2020-02-23)
Foodborne illnesses affect the health of consumers worldwide, and thus searching for potential antimicrobial agents against foodborne pathogens is given an increased focus. This research evaluated the influence of sodium lactate (SL), encapsulated (e) and unencapsulated (u) polyphosphates (PP; sodium

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service