Skip to Content
Merck
All Photos(1)

Documents

1.03857

Millipore

GranuCult® prime Levine Eosin-Methylene Blue (L-EMB) Agar

ISO 21150, acc. FDA-BAM

Synonym(s):

EMB agar, LEVINE, Eosin Methylene-blue Lactose Agar acc. to LEVINE

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106213
NACRES:
NA.74

Agency

ISO 21150

Quality Level

form

powder

technique(s)

microbiological culture: suitable

pH

7.1±0.2 (25 °C)

application(s)

food and beverages
pharmaceutical

storage temp.

15-25°C

suitability

selective and differential for enterobacteriaceae

Related Categories

General description

GranuCult® prime Eosin Methylene Blue (L-EMB) Agar is a nutrient rich medium which is designed for the isolation, confirmation, and identification of Escherichia coli, and other Enterobacteriaceae from the food chain, cosmetic and other materials.
The synergistic effect of lactose and two indicator dyes, Eosin Y and Methylene Blue differentiates colonies of lactose fermenting and non-fermenting microbes. Growing colonies of E. coli produce a green metallic sheen due to high acid production from lactose. Other coliforms do not produce enough acid to attain the metallic sheen. The growing coliforms produce blue-black colonies due to the formation of eosin-methylene blue dye complex when the pH drops.
Peptones in the medium provide nitrogen, amino acids, and minerals to support growth. Di-potassium phosphate acts as a buffer and agar is the solidifying agent.

Application

GranuCult® prime Eosin Methylene Blue (L-EMB) Agar is used for the isolation, confirmation, and identification of Escherichia coli , and other Enterobacteriaceae from the food chain, cosmetic and other materials.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service