Skip to Content
Merck
All Photos(1)

Key Documents

1.03024

Supelco

Chlorophenol red

pH indicator

Synonym(s):

Chlorophenol red, 3,3′′-Dichlorophenolsulfonphthalein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H12Cl2O5S
CAS Number:
Molecular Weight:
423.27
MDL number:
UNSPSC Code:
12352106
EC Index Number:
224-619-2

product name

Chlorophenol red, indicator

Quality Level

form

solid

loss

≤15% loss on drying, 110°C

visual transition interval

4.8-6.4, brownish-yellow to red-violet

mp

261 °C

bulk density

720 kg/m3

Amax

432-436 nm (buffer pH 4.8)
574-576 nm (buffer pH 6.4)

storage temp.

2-30°C

InChI

1S/C19H12Cl2O5S/c20-14-9-11(5-7-16(14)22)19(12-6-8-17(23)15(21)10-12)13-3-1-2-4-18(13)27(24,25)26-19/h1-10,22-23H

InChI key

WWAABJGNHFGXSJ-UHFFFAOYSA-N

Analysis Note

Identity (UV/VIS-Spectrum): passes test
Transition range: pH 4.8 - pH 6.4 brownish-yellow - red-violet
Absorption maximum λ 1 (buffer pH 4.8): 432 - 436 nm
Absorption maximum λ 2 (buffer pH 6.4): 574 - 576 nm
Spec. Absorptivity A 1%/1cm (λ1max; 0.01g/l; buffer pH 4.8; calculated on dried substance): 450 - 600
Spec. Absorptivity A 1%/1cm (λ2max; 0.01g/l; buffer pH 6.4; calculated on dried substance): 1000 - 1300
Loss on drying (110 °C): ≤ 15 %

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service