Skip to Content
Merck
All Photos(2)

Key Documents

1.02830

Supelco

Cyclohexane

for denaturation

Synonym(s):

Hexahydrobenzene, Hexamethylene, Naphthene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Beilstein:
1900225
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-806-2

vapor density

2.9 (vs air)

Quality Level

vapor pressure

168.8 mmHg ( 37.7 °C)
77 mmHg ( 20 °C)

Assay

≥99% (GC)

form

liquid

autoignition temp.

500 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

9 %

refractive index

n20/D 1.426 (lit.)

kinematic viscosity

1.26 cSt(20 °C)

bp

80.7 °C (lit.)

mp

4-7 °C (lit.)

transition temp

flash point -18 °C

solubility

0.05 g/L

density

0.779 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

C1CCCCC1

InChI

1S/C6H12/c1-2-4-6-5-3-1/h1-6H2

InChI key

XDTMQSROBMDMFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Purity (GC): ≥ 99.0 %
Identity (IR): passes test
Density (d 20 °C/ 4 °C): 0.778 - 0.779
Cyclohexane has been tested as denaturant by authorities.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service