Skip to Content
Merck
All Photos(1)

Key Documents

1.02333

Supelco

Chlorotrimethylsilane

for gas chromatography

Synonym(s):

Chlorotrimethylsilane, Trimethylchlorosilane, Trimethylsilyl chloride, TMCS

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€60.20

€60.20


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€60.20

About This Item

Linear Formula:
(CH3)3SiCl
CAS Number:
Molecular Weight:
108.64
MDL number:
UNSPSC Code:
12352103
EC Index Number:
200-900-5
NACRES:
NA.05

€60.20


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

250 hPa ( 20 °C)

Quality Level

form

liquid

autoignition temp.

420 °C

potency

4868 mg/kg LD50, oral (Rat)
1530 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5-46 % (v/v)

technique(s)

gas chromatography (GC): suitable

bp

57 °C/1013 hPa

mp

-58 °C

transition temp

flash point -20 °C

density

0.86 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

[Si](Cl)(C)(C)C

InChI

1S/C3H9ClSi/c1-5(2,3)4/h1-3H3

InChI key

IJOOHPMOJXWVHK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-18.4 °F - closed cup

Flash Point(C)

-28 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service