Skip to Content
Merck
All Photos(2)

Key Documents

1.02059

Supelco

Calcium carbonate

99.95 Suprapur®

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€296.00
500 G
€1,760.00

€296.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€296.00
500 G
€1,760.00

About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
MDL number:
UNSPSC Code:
12352302
EC Index Number:
207-439-9
NACRES:
NA.21

€296.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

description

for inorganic trace analysis

Assay

99.95%

form

solid

potency

6450 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

pH

9.5-10.5 (20 °C, 100 g/L in H2O, slurry)

mp

825 °C (decomposition)

solubility

0.017 g/L

density

2.93 g/mL at 25 °C (lit.)

bulk density

300‑1400 kg/m3

storage temp.

2-30°C

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Elastic constants of biogenic calcium carbonate.: This study investigates the elastic properties of biogenic calcium carbonate, providing valuable insights into its mechanical behavior and potential applications in biomaterials and environmental sciences (Best et al., 2024).

  • Modification of Living Diatom, Thalassiosira weissflogii, with a Calcium Precursor through a Calcium Uptake Mechanism: A Next Generation Biomaterial for Advanced Delivery Systems.: This study explores the modification of living diatoms with calcium carbonate precursors, proposing innovative applications in advanced drug delivery systems (Abdul Rahman et al., 2024).


Analysis Note

Purity (metallic): ≥ 99.95 %
Assay (complexometric): ≥ 99.0 %
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 10 ppm
Sulfate (SO₄): ≤ 20 ppm
Al (Aluminium): ≤ 0.20 ppm
Ba (Barium): ≤ 2.0 ppm
Cd (Cadmium): ≤ 0.010 ppm
Ce (Cerium): ≤ 0.050 ppm
Co (Cobalt): ≤ 0.010 ppm
Cr (Chromium): ≤ 0.050 ppm
Cu (Copper): ≤ 0.050 ppm
Eu (Europium): ≤ 0.010 ppm
Fe (Iron): ≤ 0.100 ppm
Hg (Mercury): ≤ 0.050 ppm
K (Potassium): ≤ 2.0 ppm
La (Lanthanum): ≤ 0.050 ppm
Li (Lithium): ≤ 1.0 ppm
Mg (Magnesium): ≤ 5.0 ppm
Mn (Manganese): ≤ 0.050 ppm
Na (Sodium): ≤ 3.0 ppm
Ni (Nickel): ≤ 0.050 ppm
Pb (Lead): ≤ 0.050 ppm
Rb (Rubidium): ≤ 20 ppm
Sc (Scandium): ≤ 0.010 ppm
Sm (Samarium): ≤ 0.010 ppm
Sr (Strontium): ≤ 100 ppm
Tl (Thallium): ≤ 0.01 ppm
Y (Yttrium): ≤ 0.010 ppm
Yb (Ytterbium): ≤ 0.010 ppm
Zn (Zinc): ≤ 0.10 ppm

Legal Information

SUPRAPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service