Skip to Content
Merck
All Photos(1)

Key Documents

1.01841

Supelco

Oxalic acid bis (cyclohexylidene hydrazide)

GR for analysis (reagent for copper)

Synonym(s):

Oxalic acid bis (cyclohexylidene hydrazide), Biscyclohexanone oxalyldihydrazone, Cuprizone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N4O2
CAS Number:
Molecular Weight:
278.35
MDL number:
UNSPSC Code:
12352127
EC Index Number:
206-729-2

Quality Level

form

solid

mp

208-212 °C

bulk density

150 kg/m3

storage temp.

15-25°C

InChI

1S/C14H22N4O2/c19-13(17-15-11-7-3-1-4-8-11)14(20)18-16-12-9-5-2-6-10-12/h1-10H2,(H,17,19)(H,18,20)

InChI key

DSRJIHMZAQEUJV-UHFFFAOYSA-N

Analysis Note

Assay (perchloric acid titration): ≥ 99 %
Identity (IR-spectrum): conforms
Melting range (lower value): ≥ 208 °C
Melting range (upper value): ≤ 214 °C
Suitability for copper determination: conforms

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service