Skip to Content
Merck
All Photos(1)

Key Documents

1.00606

Supelco

Iodine Test

photometric, 0.050-10.00 mg/L (I2), Spectroquant®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116116

product line

Spectroquant®

Quality Level

measuring range

0.050-10.00 mg/L (I2)

technique(s)

photometry: suitable

detection method

photometric (SPADNS - arsenic free)

storage temp.

15-25°C

General description

Method: photometric, DPD 0.050 - 10.00 mg/l I₂ Spectroquant®

Legal Information

SPECTROQUANT is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service