Skip to Content
Merck
All Photos(1)

Key Documents

1.00497

Sigma-Aldrich

AFB-Color modified

Staining kit for the detection of acid-fast bacteria (AFB) by hot staining method

Sign Into View Organizational & Contract Pricing

Select a Size

1 UNIT
€180.00

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 UNIT
€180.00

About This Item

UNSPSC Code:
41116130
NACRES:
NA.74

€180.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

clinical testing
diagnostic assay manufacturing
hematology
histology

storage temp.

15-25°C

suitability

(Acid-fast bacteria)

General description

The AFB-Color modified - Staining kit for the detection of acid-fast bacteria (AFB) by hot staining method, is used for human-medical cell diagnosis and serves the purpose of the bacteriological investigation of sample material of human origin, for example smears of enriched bacterial cultures.
AFB-Color modified staining set uses the classical Ziehl-Neelsen hot staining procedure and can be used for staining on the staining rack. The cell wall of acid-fast bacteria has a high proportion of wax and lipids and hence absorbs dyes only very slowly. The most efficient staining method is the hot staining according to Ziehl-Neelsen. In this method, carbolfuchsin solution is applied to the specimen and heated. This heating process accelerates the rate at which the fuchsin dye is absorbed and thus the formation of the mycolate-fuchsin complex in the cell wall. Once the acid-fast bacteria have absorbed the fuchsin dye, it is virtually impossible to decolorize them again, even when they are intensively treated with a decolorizing solution such as e. g. hydrochloric acid in ethanol. Accordingly, acid-fast bacteria are termed as acid- and alcohol-fast for staining, and are stained red in the microscopic visualization. Correspondingly, all non-acid-fast microorganisms are counterstained with an appropriate dye. This AFB-Color modified staining kit uses methylene blue as counterstain.
Pretreatment of the specimens with Sputofluol dissolves the bacteria from the surrounding viscid sputum and cell material.
The package is sufficient for approx. 250 applications. This product is registered as IVD and CE marked.For more details, please see instructions for use (IFU). The IFU can be downloaded from this webpage.

Application

Used to detect acid-fast bacteria" (AFB hot staining)

Analysis Note

Suitability for microscopypasses test acid-fast bacteria red Background blue

Legal Information

SPUTOFLUOL is a trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Muta. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service