Skip to Content
Merck
All Photos(1)

Key Documents

05-312

Sigma-Aldrich

Anti-E2F-4 Antibody, clone GG22-2A6

clone GG22-2A6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GG22-2A6, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable (ChIP-seq)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... E2F4(1874)

Specificity

E2F-4; does not recognize E2F-1, -2, -3, or -5

Immunogen

GST fusion to human E2F-4

Application

Detect E2F-4 with Anti-E2F-4 Antibody, clone GG22-2A6 (Mouse Monoclonal Antibody), that has been shown to work in IP & WB.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

routinely evaluated on non-stimulated A431 cell lysate

Target description

55kDa

Physical form

200μg of protein G purified mouse IgG2b in 200μl of 0.07 M Tris-glycine, pH 7.4, 0.135 M NaCl, 0.035% sodium azide and 30% glycerol.
Format: Purified
Protein G Chromatography

Storage and Stability

1 year at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dynamic loss of H2B ubiquitylation without corresponding changes in H3K4 trimethylation during myogenic differentiation.
Vethantham, V; Yang, Y; Bowman, C; Asp, P; Lee, JH; Skalnik, DG; Dynlacht, BD
Molecular and cellular biology null
Functional interaction between E2F-4 and p130: evidence for distinct mechanisms underlying growth suppression by different retinoblastoma protein family members.
Vairo, G, et al.
Genes & Development, 9, 869-881 (1995)
Erawan Borkham-Kamphorst et al.
Journal of cell communication and signaling, 6(1), 11-25 (2011-07-13)
Nephroblastoma overexpressed gene encodes a matricellular protein (CCN3/NOV) of the CCN family, comprising CCN1 (CYR61), CCN2 (CTGF), CCN4 (WISP-1), CCN5 (WISP-2), and CCN6 (WISP-3). CCN proteins are involved in the regulation of mitosis, adhesion, apoptosis, extracellular matrix production, growth arrest
Jiahui Du et al.
Development (Cambridge, England), 148(8) (2021-03-27)
Stem cells self-renew or give rise to transit-amplifying cells (TACs) that differentiate into specific functional cell types. The fate determination of stem cells to TACs and their transition to fully differentiated progeny is precisely regulated to maintain tissue homeostasis. Arid1a
L Wu et al.
Molecular and cellular biology, 20(19), 7059-7067 (2000-09-13)
Expression of the bovine papillomavirus E2 protein in cervical carcinoma cells represses expression of integrated human papillomavirus (HPV) E6/E7 oncogenes, followed by repression of the cdc25A gene and other cellular genes required for cell cycle progression, resulting in dramatic growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service