Skip to Content
Merck
All Photos(5)

Key Documents

04-789

Sigma-Aldrich

Anti-phospho-Histone H3 (Thr11) Antibody, clone MC83, rabbit monoclonal

culture supernatant, clone MC83, Upstate®

Synonym(s):

H3T11P, Histone H3 (phospho T11)

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€536.00

€536.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
200 μL
€536.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€536.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

MC83, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

inhibition assay: suitable (peptide)
multiplexing: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr11)

Gene Information

human ... H3C1(8350)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications, which include acetylation, phosphorylation, methylation, and ubiquitin-ation, occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle. Phosphorylation at Ser10, Ser28 and Thr11 of histone H3 is tightly correlated with chromosome condensation during both mitosis and meiosis. In mammals, phosphorylation of Thr11 modification is restricted to the centromeric region while in plant cells, phosphorylation of Thr11 was distributed along the entire length of condensed chromosomes. Mitosis-specific phosphorylation of histone H3 at Thr11 is mediated by Dlk/ZIP kinase.

Specificity

Broad species cross-reactivity is expected based on sequence homology. An additional unidentified band at ~40 kDa appears in some lysates.
Recognizes histone H3 phosphorylated at Thr11, Mr 17kDa.

Immunogen

Peptide containing ...KS[pT]G..., in which pT corresponds to phospho-threonine at position 11 of human histone H3

Application

Anti-phospho-Histone H3 (Thr11) Antibody, clone MC83 is a rabbit monoclonal antibody for detection of phospho-Histone H3 (Thr11) also known as H3T11P, Histone H3 (phospho T11) & has been validated in WB, PIA, Mplex.
Immunoblot Analysis:
A 1:500-1:2000 dilution of this lot detected phosphorylated histone H3 in acid extracted proteins from mitotic HeLa cells treated with colcemid, but not unmodified recombinant histone H3 (Catalog # 14-494).

Western Blot Analysis:
A 1:5,000 dilution of a previous lot detected phosphorylated histone H3 in acid extracted proteins from mitotic HeLa cells treated with colcemid, but not unmodified recombinant histone H3 (Catalog # 14-494).


Multiplexing:
Beadlyte Histone-Peptide Specificity Assay: 1:1,000-1:50,000 dilutions of a previous lot were incubated with histone H3 peptides containing various modifications conjugated to Luminex microspheres. Only the peptide containing phospho-threonine 11 was
detected (Figure C).

Peptide Inhibition Analysis: 1 μM of histone H3 peptide containing phospho-threonine 11 abolished detection of histone H3 by anti-phospho-Histone H3 (Thr11), clone MC83 in immunoblot analysis of acid extracts from
colcemid treated HeLa cells (Figure D, lane 4).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Routinely evaluated by immunoblot analysis.

Target description

17kDa

Linkage

Replaces: MABE468

Physical form

Cultured supernantant in 0.05% sodium azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt.
For maximum recovery of product, centrifuge the vial prior to removing the cap.

Analysis Note

Control
HeLa acid extracts

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan Kniewel et al.
Genetics, 207(4), 1313-1333 (2017-10-08)
Saccharomyces cerevisiae Mek1 is a CHK2/Rad53-family kinase that regulates meiotic recombination and progression upon its activation in response to DNA double-strand breaks (DSBs). The full catalog of direct Mek1 phosphorylation targets remains unknown. Here, we show that phosphorylation of histone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service