Skip to Content
Merck
All Photos(1)

Key Documents

03-188

Sigma-Aldrich

p300 (catalytic domain) human recombinant

Human recombinant p300 (catalytic domain) expressed in E. Coli.

Synonym(s):

Human recombinant p300 (catalytic domain) expressed in E. Coli, Upstate® human recombinant p300 (catalytic domain), p300 (catalytic domain) expressed in E. Coli

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€650.00

€650.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€650.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.54

€650.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

Escherichia coli

Quality Level

Assay

>75% (SDS-PAGE)

mol wt

~52 kDa

manufacturer/tradename

Upstate®

technique(s)

enzyme immunoassay: suitable

General description

Human recombinant p300 (catalytic domain) expressed in E. Coli
Although originally termed histone acetyltransferases (HATs) for their lysine acetylation activity on histone N-terminal tails, p300 and its paralogue, CBP, have been shown to acetylate a variety of non-histone proteins including p53, DNA polymerase ß and nuclear import factors. p300/CBP acetylations play regulatory roles in transcription, DNA repair and replication, the cell cycle, p53 turnover, and nuclear import. In addition to functions which overlap with those of CBP, p300 has a number of unique functions, including a role in the induction of p21Waf1/Cip1 cell cycle inhibitor.

Specificity

>250 pmol/min/µg assayed as production of CoA-SH from AcCoA in the presence of a peptide comprising human p53 residues 368-386. CoA-SH is determined colorimetrically by reaction with DTNB (5,5′-Dithiobis (2-nitrobenzoic acid).

Application

Human recombinant p300 (catalytic domain) expressed in E. Coli.

Acetyltransferase assays
Enzyme Assays

Physical form

Format: Purified
In liquid containing 50 mM Tris/HCl, pH 8.0, 0.1 mM EDTA, 10% glycerol. Frozen solution.

Storage and Stability

Stable for 12 months at -80ºC from date of receipt. Avoid multiple freeze/thaw cycles.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service