MAT089755823
Matrix Scientific
1-(5-Chloro-2-methoxy-benzenesulfonyl)-piperazine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-[(5-chloro-2-methoxyphenyl)sulfonyl]piperazine
Inchi Code
1S/C11H15ClN2O3S/c1-17-10-3-2-9(12)8-11(10)18(15,16)14-6-4-13-5-7-14/h2-3,8,13H,4-7H2,1H3
InChI key
AROSEUNOEYMHLM-UHFFFAOYSA-N
Country of Origin
US
Product Link
https://www.matrixscientific.com/018641.html
MSDS
https://www.matrixscientific.com/018641.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.