Skip to Content
Merck
All Photos(1)

Key Documents

ENAH97BD1529

Enamine

6-bromo-2-methoxy-1-nitronaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
282.09
MDL number:

mp

152-153

Iupac Name

6-bromo-2-methoxy-1-nitronaphthalene

Inchi Code

1S/C11H8BrNO3/c1-16-10-5-2-7-6-8(12)3-4-9(7)11(10)13(14)15/h2-6H,1H3

InChI key

VQUIAHIZENASND-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-7416255

Melting Point

152-153

MSDS

https://www.enaminestore.com/catalog/EN300-7416255


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library