ENAH93E743BF
Enamine
ethyl 3-[(2-bromopyridin-4-yl)oxy]-2-acetamidopropanoate
About This Item
Iupac Name
ethyl N-acetyl-O-(2-bromopyridin-4-yl)serinate
Inchi Code
1S/C12H15BrN2O4/c1-3-18-12(17)10(15-8(2)16)7-19-9-4-5-14-11(13)6-9/h4-6,10H,3,7H2,1-2H3,(H,15,16)
InChI key
GGUXGHNNOCTWJZ-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-7462564
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.