ENAH0425AAAC
Enamine
1-methyl-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carbaldehyde
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
1-methyl-5-(trifluoromethyl)-1H-1,2,3-triazole-4-carbaldehyde
Inchi Code
1S/C5H4F3N3O/c1-11-4(5(6,7)8)3(2-12)9-10-11/h2H,1H3
InChI key
GXPCQHOUNXZISH-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Ice Pack
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-1595073
MSDS
https://www.enaminestore.com/catalog/EN300-1595073
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service