Iupac Name
1-(oxiran-2-yl)cyclopentan-1-ol
Inchi Code
1S/C7H12O2/c8-7(6-5-9-6)3-1-2-4-7/h6,8H,1-5H2
InChI key
DDGASQGDKHNCQV-UHFFFAOYSA-N
storage temp.
4
Purity
95
Country of Origin
UA
Shipping Temperature
Normal
Physical Form
liquid
Product Link
https://www.enaminestore.com/catalog/EN300-344194
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-344194
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.