CIAH987EF2F7
ChemScene LLC (Preferred Partner)
5,8-Dioxa-10-azadispiro[2.0.4.3]undecane
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
bp
258.6+-25.0 C at 760 mmHg
Iupac Name
5,8-dioxa-10-azadispiro[2.0.4.3]undecane
Inchi Code
1S/C8H13NO2/c1-2-7(1)5-9-6-8(7)10-3-4-11-8/h9H,1-6H2
InChI key
IVJLQSOJUNXMDA-UHFFFAOYSA-N
storage temp.
4C
Purity
97%
Country of Origin
CN
Physical Form
Liquid
Product Link
https://www.chemscene.com/129321-60-4.html
Boiling point
258.6+-25.0 C at 760 mmHg
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://file.chemscene.com/pdf/UsaMSDS/MSDSUSACS-W007265.pdf
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.