Skip to Content
Merck
All Photos(1)

Documents

CH4H53DF6BD9

ChemBridge Corporation

([1,3]oxazolo[4,5-b]pyridin-2-ylmethyl)amine tris(trifluoroacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
491.22
MDL number:

Iupac Name

oxazolo[4,5-b]pyridin-2-ylmethanamine tris(2,2,2-trifluoroacetate)

Inchi Code

1S/C7H7N3O.3C2HF3O2/c8-4-6-10-7-5(11-6)2-1-3-9-7;3*3-2(4,5)1(6)7/h1-3H,4,8H2;3*(H,6,7)

InChI key

IIEGRKVTVXZHEK-UHFFFAOYSA-N

storage temp.

RT

Salt Data

3CF3COOH

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/4110440

MSDS

https://www.hit2lead.com/sds/4110440_sds.pdf


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service