Skip to Content
Merck
All Photos(1)

Documents

T-102

Supelco

Testosterone Calibrator

Level 7, 150 (ng/dL, Stripped Serum), pkg of 1 mL, Cerilliant®

Synonym(s):

Testosterone Calibrator

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H28O2
Molecular Weight:
288.42
UNSPSC Code:
12352200

grade

certified reference material

packaging

pkg of 1 mL

manufacturer/tradename

Cerilliant®

IVD

for in vitro diagnostic use

concentration

150 (ng/dL, Stripped Serum)

format

single component solution

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Certified reference material of testosterone in stripped serum, CE marked for in vitro diagnostic (IVD) applications by mass spectrometry. Individual levels of testosterone in stripped serum are offered separately at concentrations ranging from 2 to 2,000 ng/dL. Each individual level is ready to use as a calibrator in LC-MS/MS testosterone methods. Separate options for a blank and 13C- or deuterium labeled internal standards of testosterone or the whole kit including calibrators, blank and Testosterone-2,3,4-13C3 in 10 μg/mL Acetonitrile are also available.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service