Skip to Content
Merck
All Photos(1)

Key Documents

N-144

Supelco

Normorphine-D3 solution

100 μg/mL in methanol, certified reference material, ampule of 1 mL, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14D3NO3
Molecular Weight:
274.33
UNSPSC Code:
12352210

grade

certified reference material

Quality Level

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in methanol

format

single component solution

storage temp.

2-8°C

General description

This stable-labeled internal standard is suited for quantitation of normorphine by LC/MS or GC/MS applications such as pain prescription monitoring, urine drug testing, clinical toxicology, or forensic analysis. Normorphine is a primary urinary metabolite of the opiate analgesic morphine. Morphine, used to relieve both acute and chronic severe pain is known under trade names MS Contin®, Kadian®, Oramorph®, and Roxanol.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Kadian is a registered trademark of Actavis Elizabeth LLC
MS Contin is a registered trademark of Purdue Frederick Company
Oramorph is a registered trademark of AAIPharma, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service