Skip to Content
Merck
All Photos(1)

Documents

C-139

Supelco

Cyclosporin A-15N11

100 μg/mL in acetonitrile, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C62H11115N11O12
Molecular Weight:
1213.54
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

100 μg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−70°C

General description

A stable-labeled internal standard of cyclosporin A, an immunosuppressant and narrow therapeutic index drug used to prevent rejection in organ transplantation. This internal standard is suitable for critical quantitative applications in clinical and diagnostic testing such as therapeutic drug monitoring assays to ensure patients remain within the drug′s narrow therapeutic range. The Snap-N-Spike® format allows laboratories to eliminate weighing operations of hazardous substances such as cyclosporin A, significantly reducing occupational exposure hazards related to handling of acutely toxic powders and provides the ability to spike into their matrix of choice before use.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service