Skip to Content
Merck
All Photos(1)

Key Documents

C-124

Supelco

4-Chloro-2,5-Dimethoxyphenethylamine hydrochloride solution

1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®

Synonym(s):

2C-C hydrochloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14ClNO2 · HCl
CAS Number:
Molecular Weight:
252.14
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

1.0 mg/mL in methanol (as free base)

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology

format

single component solution

storage temp.

−20°C

InChI

1S/C10H14ClNO2.ClH/c1-13-9-6-8(11)10(14-2)5-7(9)3-4-12;/h5-6H,3-4,12H2,1-2H3;1H

InChI key

BKHKVQPFRTVWJY-UHFFFAOYSA-N

General description

2C-C, or 4-Chloro-2,5-Dimethoxyphenethylamine, is a psychedelic drug and phenethylamine of the 2C series. Sold under the street names 2C-C or 2CC the designer drug produces vivid visual hallucinations and unique among other 2C compounds, produces intense relaxing, almost sedating effects. This certified solution standard is suitable as a starting material for calibrators or controls in phenethylamine testing methods by GC/MS or LC/MS for applications in clinical toxicology, forensic analysis, or urine drug testing.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service